Tisk…

Bromoacetyl bromide ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A19580.14 A19580.22 A19580.36
A19580.14EA 1060 CZK
A19580.14 A19580.22 A19580.36
Bromoacetyl bromide ≥98%
Bromoacetyl bromide

Purity: 98% CAS No.: 598-21-0 Molecular Formula: BrCH2COBr Formula Weight: 201.86 Flash Point: 105°(221°F) Refractive Index: 1.5460 Odour: Acrid Boiling Point: 147-150° Beilstein Reference: 605440 UN No.: 2513 MDL No.: MFCD00000115

Vzorec: BrCH₂COOBr
Bod varu: 147…150 °C
Hustota: 2,324
Bod vzplanutí: >105 °C (221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000115
CAS číslo: 598-21-0
EINECS: 209-923-5
UN: 2513
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR