Bromopyruvic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid
16490-10G
16490-500G
16490-50G
SIAL16490-10GEA
3220
CZK
SIAL16490-10G
SIAL16490-500G
SIAL16490-50G
Bromopyruvic acid, Sigma-Aldrich®
Bromopyruvic acid
Vzorec:
BrH₂CC(O)COOH Bod varu: 249 °C (1013 hPa) Bod tání: 77…79 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002587 CAS číslo: 1113-59-3 EINECS: 214-206-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...