Tisk…

Bromopyruvic acid ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid

L00720.04 L00720.09 L00720.18
L00720.04EA 961 CZK
L00720.04 L00720.09 L00720.18
Bromopyruvic acid ≥97%
Bromopyruvic acid

Purity: 97% CAS No.: 1113-59-3 Molecular Formula: C3H3BrO3 Formula Weight: 166.96 Storage Temperature: 2°C to 8°C Melting Point: 75-80° Beilstein Reference: 1746786 UN No.: 3261 MDL No.: MFCD00002587

Vzorec: BrH₂CC(O)COOH
MW: 166,96 g/mol
Bod varu: 249 °C (1013 hPa)
Bod tání: 75…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002587
CAS číslo: 1113-59-3
EINECS: 214-206-5
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR