Tisk…

Ethyl bromopyruvate 80-85%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

118150250 118151000
ACRO118150250EA 2630 CZK
ACRO118150250 ACRO118151000
Ethyl bromopyruvate 80-85%
Ethyl bromopyruvate
Vzorec: BrH₂CC(O)COOCH₂CH₃
Bod varu: 100…102 °C (15 mmHg)
Hustota: 1,55 g/cm³
Bod vzplanutí: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000204
CAS číslo: 70-23-5
EINECS: 200-729-6
UN: 3265
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to pink-red Liquid
Infrared spectrum Conforms
GC ≥80 %
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Sodium carbonate (Na2CO3) ≤1.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤10 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Ag) ≤20 ppm
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR