3-Bromobutanone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
662747-1G
662747-5G
SIAL662747-1GEA
2390
CZK
SIAL662747-1G
SIAL662747-5G
3-Bromobutanone, Sigma-Aldrich®
3-Bromobutanone
Vzorec:
H₃CC(O)BrCHCH₃ Bod varu: 36 °C (1013 hPa) Hustota: 1,441 g/cm³ (18 °C) |
MDL Number:
MFCD00013538 CAS číslo: 814-75-5 EINECS: 212-404-6 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...