1-Bromo-3-chloropropane, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
B62404-1KG
B62404-250G
SIALB62404-1KGEA
3120
CZK
SIALB62404-1KG
SIALB62404-250G
1-Bromo-3-chloropropane, Sigma-Aldrich®
1-Bromo-3-chloropropane
Vzorec:
BrH₂CCH₂CH₂Cl MW: 157,44 g/mol Bod varu: 144 °C (1013 hPa) Bod tání: –58 °C Hustota: 1,6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000998 CAS číslo: 109-70-6 EINECS: 203-697-1 UN: 2688 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...