1-Bromo-2-chloroethane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ethylene chlorobromide
M-Clarity™ quality level = MQ200
232750-250G
232750-5G
SIAL232750-250GEA
3380
CZK
SIAL232750-250G
SIAL232750-5G
1-Bromo-2-chloroethane, Sigma-Aldrich®
1-Bromo-2-chloroethane
Vzorec:
BrH₂CCH₂Cl MW: 143,41 g/mol Bod varu: 107 °C (1013 hPa) Bod tání: –17 °C Hustota: 1,7392 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000962 CAS číslo: 107-04-0 EINECS: 203-456-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...