1-Bromononane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Nonyl bromide
M-Clarity™ quality level = MQ100
B74607-100G
B74607-25G
SIALB74607-100GEA
3470
CZK
SIALB74607-100G
SIALB74607-25G
1-Bromononane, Sigma-Aldrich®
1-Bromononane
Vzorec:
BrH₂C(CH₂)₇CH₃ MW: 207.15 Bod varu: 201 Bod tání: –29 °C Hustota: 1.086 Storage Temperature: Ambient |
MDL Number:
MFCD00000278 CAS číslo: 693-58-3 EINECS: 211-755-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...