Tisk…

1,12-Dibromododecane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

174866-25G 174866-5G
SIAL174866-25GEA 2650 CZK
SIAL174866-25G SIAL174866-5G
1,12-Dibromododecane, Sigma-Aldrich®
1,12-Dibromododecane
Vzorec: BrH₂C(CH₂)₁₀CH₂Br
MW: 328,13 g/mol
Bod varu: 363 °C (1013 hPa)
Bod tání: 37…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00000226
CAS číslo: 3344-70-5
EINECS: 222-096-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR