Tisk…

1,1,2,2-Tetrabromethan ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12943.AD A12943.AK A12943.AU
A12943.ADEA 1270 CZK
A12943.AD A12943.AK A12943.AU
1,1,2,2-Tetrabromethan ≥97%
1,1,2,2-Tetrabromoethane

Purity: 97% CAS No.: 79-27-6 Molecular Formula: Br2CHCHBr2 Formula Weight: 345.67 Refractive Index: 1.6370 Boiling Point: 118-120°/15mm Merck Reference: 14,9185 UN No.: 2504 MDL No.: MFCD00000133

Vzorec: Br₂HCCHBr₂
Bod varu: 118…120 °C (15 mmHg)
Bod tání: –1 °C
Hustota: 2,967
Storage Temperature: Ambient
MDL Number: MFCD00000133
CAS číslo: 79-27-6
EINECS: 201-191-5
UN: 2504
ADR: 6.1,III
Merck Index: 14,9185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecc5c2f2399032fc65a1f90353ec556de6f2f655&vid=602784cfe0c9c19c147c1108e11d03d302f55362&prid=web&clid=SAPDAM

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR