Tisk…

Ethyl sorbate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate

M-Clarity™ quality level = MQ300

W245909-100G-K W245909-1KG-K W245909-5KG-K
SIALW245909-100G-KEA 2160 CZK
SIALW245909-100G-K SIALW245909-1KG-K SIALW245909-5KG-K
Ethyl sorbate, Sigma-Aldrich®
Ethyl sorbate
Vzorec: C₈H₁₂O₂
MW: 140.18
Bod varu: 195.4
Hustota: 0.9405
MDL Number: MFCD00009296
CAS číslo: 2396-84-1
EINECS: 219-258-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR