Ethyl linoleate
Dodavatel: Cayman Chemical
10008198-100
10008198-50
10008198-500
10008198-1
CAYM10008198-100EA
828
CZK
CAYM10008198-100
CAYM10008198-50
CAYM10008198-500
CAYM10008198-1
Ethyl linoleate
Ethyl linoleate
Vzorec:
C₂₀H₃₆O₂ Storage Temperature: Freezer |
MDL Number:
MFCD00009535 CAS číslo: 544-35-4 EINECS: 208-868-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...