Batofenantrolin, Supelco®
Dodavatel: Merck
Synonyms:
4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
M-Clarity™ quality level = MQ100
11880-1G-F
11880-5G-F
11880-1G-FEA
4550
CZK
11880-1G-F
11880-5G-F
Batofenantrolin, Supelco®
Batofenantrolin
Vzorec:
C₂₄H₁₆N₂ MW: 332,4 g/mol Bod varu: 508 °C (1013 hPa) Bod tání: 215…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004976 CAS číslo: 1662-01-7 EINECS: 216-767-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...