Tisk…

Batofenantrolin, bílý, powder

Dodavatel: MP Biomedicals

Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)

02190157.5 0219015701
ICNA02190157.5EA 3620 CZK
ICNA02190157.5 ICNA0219015701
Batofenantrolin, bílý, powder
Batofenantrolin

Bathophenanthroline is used as a bidentate ligand and for the colorimetric determination of iron.


Bathophenanthroline is used as a bidentate ligand and for the colorimetric determination of iron.


Soluble in 95% ethanol:chloroform (2:1) (50 mg/ml. Insoluble in water.

Vzorec: C₂₄H₁₆N₂
MW: 332,4 g/mol
Bod varu: 508 °C (1013 hPa)
Bod tání: 215…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00004976
CAS číslo: 1662-01-7
EINECS: 216-767-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR