Tisk…

3,4,7,8-Tetramethyl-1,10-phenanthroline ≥98%

Dodavatel: Thermo Fisher Scientific

L01482.03 L01482.06
L01482.03EA 3200 CZK
L01482.03 L01482.06
3,4,7,8-Tetramethyl-1,10-phenanthroline ≥98%
3,4,7,8-Tetramethyl-1,10-phenanthroline

Purity: 98+% CAS No.: 1660-93-1 Molecular Formula: C16H16N2 Formula Weight: 236.32 Melting Point: ca 280° dec. Beilstein Reference: 171279

Vzorec: C₁₆H₁₆N₂
Bod varu: 386 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00004974
CAS číslo: 1660-93-1
EINECS: 216-762-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR