Tisk…

5-Nitro-1,10-phenanthroline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

N8501-1G N8501-5G
SIALN8501-1GEA 4150 CZK
SIALN8501-1G SIALN8501-5G
5-Nitro-1,10-phenanthroline, Sigma-Aldrich®
5-Nitro-1,10-phenanthroline
Vzorec: C₁₂H₇N₃O₂
Bod tání: 203 °C
MDL Number: MFCD00004981
CAS číslo: 4199-88-6
EINECS: 224-097-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR