Tisk…

5-Nitro-1,10-phenanthroline ≥98%

Dodavatel: Thermo Scientific
Varování

A11428.03 A11428.06 A11428.14
A11428.03EA 1920 CZK
A11428.03 A11428.06 A11428.14
5-Nitro-1,10-phenanthroline ≥98%
5-Nitro-1,10-phenanthroline

Purity: 98% CAS No.: 4199-88-6 Molecular Formula: C12H7N3O2 Formula Weight: 225.21 Melting Point: 202-204° Beilstein Reference: 196245

Vzorec: C₁₂H₇N₃O₂
Bod tání: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00004981
CAS číslo: 4199-88-6
EINECS: 224-097-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR