Tisk…

D-Glukono-1,5-lakton, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone

M-Clarity™ quality level = MQ200

G4750-100G G4750-1KG G4750-500G
SIALG4750-100GEA 2090 CZK
SIALG4750-100G SIALG4750-1KG SIALG4750-500G
D-Glukono-1,5-lakton, Sigma-Aldrich®
D-Glukono-1,5-lakton
Vzorec: C₆H₁₀O₆
MW: 178.14
Bod varu: 385
Bod tání: 153
Hustota: 1.61 g/cm³ (–5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS číslo: 90-80-2
EINECS: 202-016-5
Merck Index: 13,04470

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR