D(+)-Biotin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
M-Clarity™ quality level = MQ200
B0301-25G
B0301-500MG
SIALB0301-25GEA
0
CZK
SIALB0301-25G
SIALB0301-500MG
D(+)-Biotin, Sigma-Aldrich®
D(+)-Biotin
Vzorec:
C₁₀H₁₆N₂O₃S MW: 244.31 Bod tání: 232…233 °C Hustota: 1.41 Storage Temperature: Refrigerator |
MDL Number:
MFCD00005541 CAS číslo: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...