(S)-(-)-Limonene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
M-Clarity™ quality level = MQ300
W504505-1KG-K
W504505-8KG-K
SIALW504505-1KG-KEA
3280
CZK
SIALW504505-1KG-K
SIALW504505-8KG-K
(S)-(-)-Limonene, Sigma-Aldrich®
(S)-(-)-Limonene
Vzorec:
C₁₀H₁₆ MW: 136.24 Bod varu: 175 Bod tání: –74 °C Hustota: 0.84 g/cm³ (20 °C) Bod vzplanutí: 43 Storage Temperature: Ambient |
MDL Number:
MFCD00001558 CAS číslo: 5989-54-8 EINECS: 227-815-6 UN: 2052 ADR: 3 III Merck Index: 13,05515 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...