(1S,2S)-(+)-1,2-Diaminocyclohexane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(S,S)-DACH, (1S,2S)-Cyclohexane-1,2-diamine
M-Clarity™ quality level = MQ100
346713-1G
346713-5G
SIAL346713-1GEA
2840
CZK
SIAL346713-1G
SIAL346713-5G
(1S,2S)-(+)-1,2-Diaminocyclohexane, Sigma-Aldrich®
(1S,2S)-(+)-1,2-Diaminocyclohexane
Vzorec:
C₆H₁₄N₂ MW: 114,19 g/mol Bod varu: 187 °C (1013 hPa) Bod tání: 40…43 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00062986 CAS číslo: 21436-03-3 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...