(1R,2R)-(-)-1,2-Diaminocyclohexane 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98%
CAS No.: 20439-47-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Odour: Amine-like
Melting Point: 40-43°
Boiling Point: 104-110°/40mm
Beilstein Reference: 4780911
Optical Rotation: -25.5° (c=5 in 1N HCl)
UN No.: 3259
MDL No.: MFCD00062985
Vzorec:
C₆H₁₄N₂ Bod varu: 104…110 °C (40 mmHg) Bod tání: 40…43 °C Bod vzplanutí: 70 °C (158 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00062985 CAS číslo: 20439-47-8 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...