(S)-(-)-1-Phenylethanol, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
(S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol
M-Clarity™ quality level = MQ100
05512-25G-F
05512-5G-F
SIAL05512-25G-FEA
11820
CZK
SIAL05512-25G-F
SIAL05512-5G-F
(S)-(-)-1-Phenylethanol, Sigma-Aldrich®
(S)-(-)-1-Phenylethanol
Vzorec:
C₈H₁₀O MW: 122,17 g/mol Bod varu: 207 °C (1013 hPa) Hustota: 1,018 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064264 CAS číslo: 1445-91-6 UN: 2937 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...