(R)-(+)-1-Phenyl-1-propanol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(R)-(+)-α-Ethylbenzyl alcohol
L05681.MC
L05681.ME
L05681.MCEA
1240
CZK
L05681.MC
L05681.ME
(R)-(+)-1-Phenyl-1-propanol ≥99%
(R)-(+)-1-Phenyl-1-propanol
Purity: 99%
CAS No.: 1565-74-8
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 95°(203°F)
Refractive Index: 1.5200
Boiling Point: 217-219°
Beilstein Reference: 2041555
Optical Rotation: +47° (c=2.2 in hexane)
MDL No.: MFCD00064279
Vzorec:
C₉H₁₂O MW: 136.19 Bod varu: 217…219 °C Hustota: 0.993 Bod vzplanutí: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064279 CAS číslo: 1565-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...