Tisk…

(S)-(-)-α-Methylbenzylamine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: (S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine

M-Clarity™ quality level = MQ100

115568-100G 115568-25G
SIAL115568-100GEA 4820 CZK
SIAL115568-100G SIAL115568-25G
(S)-(-)-α-Methylbenzylamine, Sigma-Aldrich®
(S)-(-)-α-Methylbenzylamine
Vzorec: C₈H₁₁N
MW: 121,18 g/mol
Bod varu: 187 °C (1013 hPa)
Bod tání: –10 °C
Hustota: 0,95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064406
CAS číslo: 2627-86-3
EINECS: 220-098-0
Merck Index: 12,06106

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR