(R)-(+)-α-Methylbenzylamine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
M-Clarity™ quality level = MQ200
77880-10ML
77880-250ML
77880-50ML
SIAL77880-10MLEA
756
CZK
SIAL77880-10ML
SIAL77880-250ML
SIAL77880-50ML
(R)-(+)-α-Methylbenzylamine, Sigma-Aldrich®
(R)-(+)-α-Methylbenzylamine
Vzorec:
C₈H₁₁N MW: 121,18 g/mol Bod varu: 180 °C (1013 hPa) Bod tání: 32,5 °C Hustota: 0,95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064405 CAS číslo: 3886-69-9 EINECS: 223-423-4 Merck Index: 12,06106 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...