Tisk…

(R)-(+)-α-Methylbenzylamine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine

M-Clarity™ quality level = MQ200

115541-100ML 115541-25ML 115541-10ML
SIAL115541-100MLEA 6050 CZK
SIAL115541-100ML SIAL115541-25ML SIAL115541-10ML
(R)-(+)-α-Methylbenzylamine, Sigma-Aldrich®
(R)-(+)-α-Methylbenzylamine
Vzorec: C₈H₁₁N
MW: 121,18 g/mol
Bod varu: 180 °C (1013 hPa)
Bod tání: 32,5 °C
Hustota: 0,95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064405
CAS číslo: 3886-69-9
EINECS: 223-423-4
Merck Index: 12,06106

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR