Tisk…

L-Tyrosin ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

A11141.0B A11141.18 A11141.22 A11141.30
A11141.0BEA 10090 CZK
A11141.0B A11141.18 A11141.22 A11141.30
L-Tyrosin ≥99%
L-Tyrosin

Purity: 99% CAS No.: 60-18-4 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Physical Form: Cryst powder Density: 1.46 Appearance: White Melting Point: 300° Beilstein Reference: 392441 Merck Reference: 14,9839 Optical Rotation: -11° (c=4 in 1N HCl) MDL No.: MFCD00002606

Vzorec: C₉H₁₁NO₃
MW: 181.19
Bod varu: 351
Bod tání: >300 °C
Hustota: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS číslo: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR