Tisk…

(S)-(+)-Hydratropic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (S)-2-Phenylpropanoic acid, (S)-(+)-2-Phenylpropionic acid, (S)-(+)-2-Phenylpropanoic acid

M-Clarity™ quality level = MQ100

279900-1G 279900-250MG
SIAL279900-1GEA 13810 CZK
SIAL279900-1G SIAL279900-250MG
(S)-(+)-Hydratropic acid, Sigma-Aldrich®
(S)-(+)-Hydratropic acid
Vzorec: C₉H₁₀O₂
MW: 150.18
Bod varu: 257
Bod tání: 30…31 °C
Hustota: 1.1
Storage Temperature: Refrigerator
MDL Number: MFCD00063139
CAS číslo: 7782-24-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR