(S)-(+)-Hydratropic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(S)-2-Phenylpropanoic acid, (S)-(+)-2-Phenylpropionic acid, (S)-(+)-2-Phenylpropanoic acid
L08802.03
L08802.MD
L08802.03EA
2510
CZK
L08802.03
L08802.MD
(S)-(+)-Hydratropic acid ≥97%
(S)-(+)-Hydratropic acid
Purity: 97%
CAS No.: 7782-24-3
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 148°(298°F)
Refractive Index: 1.5230
Melting Point: 30-34°
Boiling Point: 264-266°
Beilstein Reference: 2044507
Optical Rotation: +73° (c=1.6 in chloroform)
MDL No.: MFCD00063139
Vzorec:
C₉H₁₀O₂ Bod varu: 264…266 °C Bod tání: 30…34 °C Hustota: 1,100 Bod vzplanutí: 148 °C (298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063139 CAS číslo: 7782-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...