Tisk…

(S)-(+)-2-Phenylbutyric acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

279889-1G 279889-5G
SIAL279889-1GEA 2710 CZK
SIAL279889-1G SIAL279889-5G
(S)-(+)-2-Phenylbutyric acid, Sigma-Aldrich®
(S)-(+)-2-Phenylbutyric acid
Vzorec: C₁₀H₁₂O₂
MW: 164,2 g/mol
MDL Number: MFCD00063164
CAS číslo: 4286-15-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR