(S)-(+)-2-Phenylbutyric acid ≥70%
Dodavatel: Thermo Scientific
|
Varování
|
L12754.03
L12754.06
L12754.03EA
3360
CZK
L12754.03
L12754.06
(S)-(+)-2-Phenylbutyric acid ≥70%
(S)-(+)-2-Phenylbutyric acid
Purity: 99%
CAS No.: 4286-15-1
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point:
Vzorec:
C₁₀H₁₂O₂ MW: 164,2 g/mol Bod varu: 270…272 °C Hustota: 1,055 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063164 CAS číslo: 4286-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...