(R)-(-)-4-Methyl-2-pentanol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L18881.03
L18881.MD
L18881.03EA
4190
CZK
L18881.03
L18881.MD
(R)-(-)-4-Methyl-2-pentanol ≥99%
(R)-(-)-4-Methyl-2-pentanol
Purity: 99%
CAS No.: 16404-54-9
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 41°(105°F)
Refractive Index: 1.4110
Odour: Slightly sweetish
Boiling Point: 130-131°
Beilstein Reference: 1718992
Optical Rotation: -21° (neat)
UN No.: 2053
MDL No.: MFCD03093077
Vzorec:
C₆H₁₄O MW: 102.18 Bod varu: 130…131 °C Hustota: 0.802 Bod vzplanutí: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03093077 CAS číslo: 16404-54-9 UN: 2053 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...