Tisk…

L(+)-Leucinol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

B23745.06 B23745.14
B23745.06EA 2360 CZK
B23745.06 B23745.14
L(+)-Leucinol ≥97%
L(+)-Leucinol

Purity: 97% CAS No.: 7533-40-6 Molecular Formula: C6H15NO Formula Weight: 117.19 Flash Point: 90°(194°F) Refractive Index: 1.4510 Boiling Point: 198-200° Beilstein Reference: 1719240 Optical Rotation: +1.1° (neat) MDL No.: MFCD00063676

Vzorec: C₆H₁₅NO
Bod varu: 198…200 °C
Bod tání: 66…70 °C
Hustota: 0,917
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063676
CAS číslo: 7533-40-6
EINECS: 231-400-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR