Tisk…

(S)-(+)-2-Amino-3-methyl-1-butanol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol

L11300.03 L11300.06 L11300.14
L11300.03EA 1080 CZK
L11300.03 L11300.06 L11300.14
(S)-(+)-2-Amino-3-methyl-1-butanol ≥97%
(S)-(+)-2-Amino-3-methyl-1-butanol

Purity: 97% CAS No.: 2026-48-4 Molecular Formula: C5H13NO Formula Weight: 103.17 Storage Temperature: 2°C to 8°C Flash Point: 78°(172°F) Refractive Index: 1.4548 Melting Point: 30-35° Boiling Point: 80-81°/8mm Beilstein Reference: 1719137 Optical Rotation: +17° (c=10 in ethanol) MDL No.: MFCD00064296

Vzorec: C₅H₁₃NO
Bod varu: 80…81 °C (8 mmHg)
Bod tání: 30…35 °C
Hustota: 0,926
Bod vzplanutí: 78 °C (172 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00064296
CAS číslo: 2026-48-4
EINECS: 217-975-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR