D-(-)-Leucinol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
298417-1G
298417-5G
SIAL298417-1GEA
2760
CZK
SIAL298417-1G
SIAL298417-5G
D-(-)-Leucinol, Sigma-Aldrich®
D-(-)-Leucinol
Vzorec:
C₆H₁₅NO Bod varu: 199 °C (1013 hPa) Hustota: 0,897 g/cm³ (20 °C) |
MDL Number:
MFCD00004734 CAS číslo: 53448-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...