Tisk…

L(+)-Leucin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

L8912-100G L8912-10MG L8912-1KG L8912-25G
SIALL8912-100GEA 7510 CZK
SIALL8912-100G SIALL8912-10MG SIALL8912-1KG SIALL8912-25G
L(+)-Leucin, Sigma-Aldrich®
L(+)-Leucin
Vzorec: C₆H₁₃NO₂
Bod tání: 300 °C
Hustota: 1,165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS číslo: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR