Tisk…

D-(-)-Leucine ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-D-Leu-OH, (R)-(-)-Leucine

A14842.06 A14842.14 AB117224.0025
A14842.06EA 1600 CZK
A14842.06 A14842.14 ABCRAB117224.0025
D-(-)-Leucine ≥99%
D-(-)-Leucine

Purity: 99% CAS No.: 328-38-1 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721721

Vzorec: C₆H₁₃NO₂
Bod varu: 226 °C (1013 hPa)
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063088
CAS číslo: 328-38-1
EINECS: 206-327-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR