Tisk…

D-(-)-Valine ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-D-Val-OH, (R)-(-)-Valine

A18894.06 A18894.14 A18894.22
A18894.06EA 1240 CZK
A18894.06 A18894.14 A18894.22
D-(-)-Valine ≥98%
D-(-)-Valine

Purity: 98+% CAS No.: 640-68-6 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: 295° dec. Beilstein Reference: 1721135

Vzorec: C₅H₁₁NO₂
MW: 117.15
Bod varu: 217
Bod tání: 295 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00064219
CAS číslo: 640-68-6
EINECS: 211-368-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR