L(+)-Izoleucin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH
M-Clarity™ quality level = MQ300
W527602-100G
W527602-1KG
SIALW527602-100GEA
2010
CZK
SIALW527602-100G
SIALW527602-1KG
L(+)-Izoleucin, Sigma-Aldrich®
L(+)-Izoleucin
Vzorec:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Bod tání: 279…280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064222 CAS číslo: 73-32-5 EINECS: 200-798-2 Merck Index: 14,05179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...