Tisk…

L(+)-Izoleucin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

I7403-100G I7403-10MG I7403-1KG I7403-25G
SIALI7403-100GEA 21970 CZK
SIALI7403-100G SIALI7403-10MG SIALI7403-1KG SIALI7403-25G
L(+)-Izoleucin, Sigma-Aldrich®
L(+)-Izoleucin
Vzorec: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Bod tání: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS číslo: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR