Tisk…

D(-)-allo-Isoleucine ≥97%

Dodavatel: Thermo Scientific

Synonyms: H-D-Allo-Ile-OH, (2R,3S)-(-)-allo-Isoleucine, H-D-allo-Ile-OH

H27189.MD H27189.06
H27189.MDEA 3380 CZK
H27189.MD H27189.06
D(-)-allo-Isoleucine ≥97%
D(-)-allo-Isoleucine

Purity: 97% CAS No.: 1509-35-9 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: ca 285° dec. Beilstein Reference: 1721794

Vzorec: C₆H₁₃NO₂
MW: 131,17 g/mol
Bod tání: ∼ 285 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00066445
CAS číslo: 1509-35-9
EINECS: 216-143-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR