D-(-)-Isoleucine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2R,3R)-(-)-Isoleucine, H-D-Ile-OH
M-Clarity™ quality level = MQ200
I7634-1G
I7634-250MG
SIALI7634-1GEA
46440
CZK
SIALI7634-1G
SIALI7634-250MG
D-(-)-Isoleucine, Sigma-Aldrich®
D-(-)-Isoleucine
Vzorec:
H₃CCH₂CH(CH₃)CH(NH₂)COOH MW: 131,17 g/mol Bod varu: 235 °C (1013 hPa) Bod tání: 98 °C Hustota: 1,224 g/cm³ (20 °C) |
MDL Number:
MFCD00064221 CAS číslo: 319-78-8 EINECS: 206-269-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...