Tisk…

(S,S)-(+)-2,4-Pentanediol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2S,4S)-2,4-Pentanediol, (S,S)-(+)-2,4-Dihydroxy pentane, (2S,4S)-(+)-Pentanediol, (2S,4S)-(+)-2,4-Pentanediol

M-Clarity™ quality level = MQ100

267872-500MG 267872-5G
SIAL267872-500MGEA 3430 CZK
SIAL267872-500MG SIAL267872-5G
(S,S)-(+)-2,4-Pentanediol, Sigma-Aldrich®
(S,S)-(+)-2,4-Pentanediol
Vzorec: C₅H₁₂O₂
MW: 104,15 g/mol
Bod varu: 111…113 °C (19 mmHg)
Bod tání: 45…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00064276
CAS číslo: 72345-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR