(S)-(+)-2-Hexanol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(S)-2-Hexanol, (S)-(+)-Butyl methyl carbinol, (S)-(+)-Methyl butyl carbinol, (S)-(+)-2-Hydroxy hexane
L10401.03
L10401.MD
L10401.03EA
4640
CZK
L10401.03
L10401.MD
(S)-(+)-2-Hexanol ≥98%
(S)-(+)-2-Hexanol
Purity: 98%
CAS No.: 52019-78-0
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 45°(113°F)
Refractive Index: 1.4140
Boiling Point: 137-138°
Beilstein Reference: 1718997
Optical Rotation: +10.5° (neat)
UN No.: 2282
MDL No.: MFCD00065955
Vzorec:
C₆H₁₄O MW: 102,18 g/mol Bod varu: 137…138 °C Hustota: 0,818 Bod vzplanutí: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00065955 CAS číslo: 52019-78-0 UN: 2282 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...