(R)-(-)-2-Octanol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol
L11502.03
L11502.06
L11502.03EA
1240
CZK
L11502.03
L11502.06
(R)-(-)-2-Octanol ≥99%
(R)-(-)-2-Octanol
Purity: 99%
CAS No.: 5978-70-1
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 76°(169°F)
Refractive Index: 1.4260
Odour: Pleasant
Boiling Point: 179-180°
Beilstein Reference: 1719324
Merck Reference: 14,6752
Optical Rotation: -10° (c=1 in ethanol)
MDL No.: MFCD00064284
Vzorec:
C₈H₁₈O MW: 130,23 g/mol Bod varu: 179…180 °C Hustota: 0,819 Bod vzplanutí: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064284 CAS číslo: 5978-70-1 EINECS: 227-777-0 UN: 1987 ADR: 3,III Merck Index: 14,6752 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...