(R)-(-)-2-Butanol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
M-Clarity™ quality level = MQ100
236691-10G
236691-1G
SIAL236691-10GEA
16630
CZK
SIAL236691-10G
SIAL236691-1G
(R)-(-)-2-Butanol, Sigma-Aldrich®
(R)-(-)-2-Butanol
Vzorec:
C₄H₁₀O MW: 74,12 g/mol Bod varu: 99 °C (1013 hPa) Bod tání: –114 °C Hustota: 0,807 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064280 CAS číslo: 14898-79-4 EINECS: 238-967-8 UN: 1120 ADR: 3,III Merck Index: 13,01540 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...