Tisk…

(S)-(+)-2-Amino-1-propanol 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol

B24916.03 B24916.06 B24916.14
B24916.03EA 1040 CZK
B24916.03 B24916.06 B24916.14
(S)-(+)-2-Amino-1-propanol 98%
(S)-(+)-2-Amino-1-propanol

Purity: 98% CAS No.: 2749-11-3 Molecular Formula: C3H9NO Formula Weight: 75.11 Flash Point: 83°(181°F) Refractive Index: 1.4498 Boiling Point: 174-176° Beilstein Reference: 1718865 Optical Rotation: +22° (c=2 in ethanol) UN No.: 2735 MDL No.: MFCD00064412

Vzorec: C₃H₉NO
MW: 75,11 g/mol
Bod varu: 174…176 °C
Hustota: 0,965
Bod vzplanutí: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS číslo: 2749-11-3
EINECS: 220-388-7
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR