(S)-(+)-2-Amino-1-butanol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol
L11449.06
L11449.03
L11449.06EA
2520
CZK
L11449.06
L11449.03
(S)-(+)-2-Amino-1-butanol ≥98%
(S)-(+)-2-Amino-1-butanol
Purity: 98+%
CAS No.: 5856-62-2
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 89°(192°F)
Refractive Index: 1.4525
Odour: Amine-like
pH: 11.1
Boiling Point: 176-178°
Beilstein Reference: 1718930
Optical Rotation: +12° (c=2 in ethanol)
UN No.: 2735
MDL No.: MFCD00064418
Vzorec:
C₄H₁₁NO Bod varu: 176…178 °C Bod tání: –2 °C Hustota: 0,947 Bod vzplanutí: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064418 CAS číslo: 5856-62-2 EINECS: 227-475-9 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...