Tisk…

(R)-(-)-2-Amino-1-propanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: D-Alaninol, D(-)-2-Amino-1-propanol, D(-)-Alaninol, (R)-2-Aminopropan-1-ol

M-Clarity™ quality level = MQ100

297682-1G 297682-5G
SIAL297682-1GEA 1310 CZK
SIAL297682-1G SIAL297682-5G
(R)-(-)-2-Amino-1-propanol, Sigma-Aldrich®
(R)-(-)-2-Amino-1-propanol
Vzorec: C₃H₉NO
MW: 75,11 g/mol
Bod varu: 173 °C (1013 hPa)
Hustota: 0,96 g/cm³ (20 °C)
MDL Number: MFCD00064413
CAS číslo: 35320-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR